Learn More
Medchemexpress LLC Posaconazole-d4 | 1133712-26-1 | 99.9% | 704.80 | 1 MG
SDP
Catalog No. 5000384952
Encompass_Preferred
Posaconazole-d4 is a deuterium-labeled form of Posaconazole, a broad-spectrum, second-generation triazole compound with antifungal activity. This compound is suitable for various laboratory applications.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-84952 Supplier Medchemexpress LLC Supplier No. HY17373S11MG
Posaconazole-d4 is a deuterium-labeled form of Posaconazole, a broad-spectrum, second-generation triazole compound with antifungal activity. This compound is suitable for various laboratory applications.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS