Learn More
Medchemexpress LLC Posaconazole-d5 | 1217785-83-5 | 99.1% | 705.81 | 1 MG
SDP
Catalog No. 5000385215
Encompass_Preferred
Posaconazole-d5 is a deuterium-labeled form of Posaconazole, a broad-spectrum triazole compound with antifungal activity. This compound is suitable for various quantitative analyses and as a tracer in research applications.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Broad-spectrum antifungal activity
Catalog No. 50-003-85215 Supplier Medchemexpress LLC Supplier No. HY17373S1MG
Posaconazole-d5 is a deuterium-labeled form of Posaconazole, a broad-spectrum triazole compound with antifungal activity. This compound is suitable for various quantitative analyses and as a tracer in research applications.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Broad-spectrum antifungal activity