Learn More
Medchemexpress LLC Posaconazole-d5 | 1217785-83-5 | 99.1% | 705.81 g/mol | C37H37D5F2N8O4 | 10 MG
SDP
Catalog No. 5000298749
Encompass_Preferred
Posaconazole-d5 is the deuterium-labeled analogue of posaconazole, provided as a high-purity stable isotope for use as an internal standard or tracer in analytical workflows. It is intended for quantitative method development and bioanalytical applications, supporting accurate quantitation in complex matrices when used with appropriate instrumentation and controls.
  • High isotopic purity for reliable internal standard performance.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • High chemical purity for reproducible analytical results.
  • Available in small-mass vials convenient for trace-level experiments.
  • Stable under recommended storage conditions to preserve integrity.
Catalog No. 50-002-98749 Supplier Medchemexpress LLC Supplier No. HY17373S10MG
Posaconazole-d5 is the deuterium-labeled analogue of posaconazole, provided as a high-purity stable isotope for use as an internal standard or tracer in analytical workflows. It is intended for quantitative method development and bioanalytical applications, supporting accurate quantitation in complex matrices when used with appropriate instrumentation and controls.
  • High isotopic purity for reliable internal standard performance.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • High chemical purity for reproducible analytical results.
  • Available in small-mass vials convenient for trace-level experiments.
  • Stable under recommended storage conditions to preserve integrity.