Learn More
Selleck Chemical LLC Posaconazole 25mg 171228-49-2 SCH56592
SDP
Catalog No. 507294
Encompass_Preferred
Posaconazole (SCH56592) is an inhibitor primarily of CYP3A4, but it does not inhibit the activity of other CYP enzymes; Also an inhibitor of sterol C14? demethylase inhibitor with IC50 of 0.25 ?M. Posaconazole has a median terminal elimination half-life of 15-35 hours. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-729-4 Supplier Selleck Chemical LLC Supplier No. S125725MG
Posaconazole (SCH56592) is an inhibitor primarily of CYP3A4, but it does not inhibit the activity of other CYP enzymes; Also an inhibitor of sterol C14? demethylase inhibitor with IC50 of 0.25 ?M. Posaconazole has a median terminal elimination half-life of 15-35 hours. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.