Learn More
TARGETMOL CHEMICALS INC POSATIRELIN 5MG
SDP
Catalog No. 502580279
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Posatirelin (BRN6006446) is a synthetic peptide with modulatory activity and neurotrophic effects on monoaminergic and cholinergic systems for the study of vascular dementia. purity: 98%
Catalog No. 50-258-0279 Supplier TARGETMOL CHEMICALS INC Supplier No. T341145MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Posatirelin (BRN6006446) is a synthetic peptide with modulatory activity and neurotrophic effects on monoaminergic and cholinergic systems for the study of vascular dementia. purity: 98%