Learn More
Access Technologies POSI-GRIP HUBER NEEDLE 22G D
Catalog No. NC9926349
Encompass
22 gauge Posigrip Huber needles 3/4 in. long with injection caps
Catalog No. NC9926349 Supplier Access Technologies Supplier No. PG2275C
22 gauge Posigrip Huber needles 3/4 in. long with injection caps