Learn More
Cell Signaling Technology Posibeads™ G4S Linker Positive Control Beads, 5 nm Polystyrene, 50 Assays
SDP
Catalog No. 503132918
Encompass
These 5 nm non-fluorescent polystyrene beads are coated with G4S linker peptide to serve as a specific positive control for confirming binding and fluorescence of G4S linker-targeted antibody conjugates in flow cytometry. The beads behave like cells in suspension and can be processed alongside cell samples using standard immunostaining and wash protocols. They may be used as a standalone sample or as an internal positive control to verify antibody activity and allow for consistent fluorescence comparisons across experiments and instruments.
  • Specific control for G4S linker-targeted antibodies
  • Functions as a standalone sample or internal positive control
  • Compatible with standard cell-based immunostaining and wash protocols
  • Enables consistent data comparison across instruments and experiments
  • Supplied in 1X PBS (pH 7.4), 0.05% Tween 20, and 0.05% sodium azide
Catalog No. 50-313-2918 Supplier Cell Signaling Technology Supplier No. 88651S
These 5 nm non-fluorescent polystyrene beads are coated with G4S linker peptide to serve as a specific positive control for confirming binding and fluorescence of G4S linker-targeted antibody conjugates in flow cytometry. The beads behave like cells in suspension and can be processed alongside cell samples using standard immunostaining and wash protocols. They may be used as a standalone sample or as an internal positive control to verify antibody activity and allow for consistent fluorescence comparisons across experiments and instruments.
  • Specific control for G4S linker-targeted antibodies
  • Functions as a standalone sample or internal positive control
  • Compatible with standard cell-based immunostaining and wash protocols
  • Enables consistent data comparison across instruments and experiments
  • Supplied in 1X PBS (pH 7.4), 0.05% Tween 20, and 0.05% sodium azide