Learn More
Andwin Scientific POSIDYNE MEM KP CAPSULE 0.1UM
SDP
Supplier:  Andwin Scientific KA2NTZP2
Encompass_Preferred
Posidyne membrane in Kleenpak capsule, 0.1m removal rating, 800 cm EFA, - in. hose barb connection
Catalog No. NC2094302