Learn More
Globe Scientific PosiStop LTS-Compatible Pipette Tips, Empty Racks
Catalog No. 23763014
Change view
Click to view available options
Holds:
Pipette Tips (1000 μL)
Pipette Tips (20 μL)
Pipette Tips (200 μL)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
23-763-014 Pipette Tips (20 μL)
23-763-015 Pipette Tips (200 μL)
23-763-016 Pipette Tips (1000 μL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-763-014 Supplier Globe Scientific Supplier No. 154405
Add to Cart
Add to Cart
This item is not returnable. View return policy
Empty racks for PosiStop pipette tips in 20 μL (red tray), 200 μL (green tray), and 1000 μL (blue tray) sizes. Durable, reusable, and color-coded for quick volume ID and efficient lab organization. Ideal for refilling and maintaining clean, organized tip storage.
  • Empty racks for 20 μL, 200 μL, and 1000 μL PosiStop pipette tips
  • Color-coded trays: red (20 μL), green (200 μL), blue (1000 μL)
  • Durable and reusable for efficient lab workflows
  • Ideal for refilling and organized tip storage
Specifications
Autoclavable Autoclavable
Color Natural
Holds Pipette Tips (20 μL)
Material Polypropylene
Positions 96
Shape Rectangular
Stackable Yes
Sterility Non-sterile
Product Type Empty Rack
Array 12 x 8