Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Positioner 8 950 mL Glass

Catalog No. 5000257087 Shop All Hach Company Products
Encompass

Retainer positioner for eight 950 ml glass bottles

Catalog No. 50-002-57087 Supplier Hach Company Supplier No. 2347
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Retainer positioner for eight 950 ml glass bottles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.