Learn More
Med Vet International Avian Positioner, 18in x 18in, J0381P, Each
Catalog No. 5000495477
Encompass_Preferred
Auspex Avian Positioner is used to position an avian species for radiography and surgery. The Positioner is made of a piece of durable plexiglass board. There are two short slots and four longer slots machined through the board. Several pieces of strategically-placed velcro hook type fasteners are permanently attached to the bottom of the board. Several straps of loop type velcro are supplied, which are utilized for restraining the bird’s and wings. Several pairs of the velcro face straps, used to restrain the bird’s legs, are also included. The versatile Auspex Avian Positioner can be applied to most types of birds, from budgerigars to macaws. It is highly effective and reliable, able to securely restrain even the most unruly bird. It is safe and relatively painless, since the bird is held using soft cloth-like straps, unlike the guillotine-type devices that hold the bird’s neck down with a piece of hard plastic. The Positioner is low-cost and durable.
Catalog No. 50-004-95477 Supplier Med Vet International Supplier No. JOR381P
Auspex Avian Positioner is used to position an avian species for radiography and surgery. The Positioner is made of a piece of durable plexiglass board. There are two short slots and four longer slots machined through the board. Several pieces of strategically-placed velcro hook type fasteners are permanently attached to the bottom of the board. Several straps of loop type velcro are supplied, which are utilized for restraining the bird’s and wings. Several pairs of the velcro face straps, used to restrain the bird’s legs, are also included. The versatile Auspex Avian Positioner can be applied to most types of birds, from budgerigars to macaws. It is highly effective and reliable, able to securely restrain even the most unruly bird. It is safe and relatively painless, since the bird is held using soft cloth-like straps, unlike the guillotine-type devices that hold the bird’s neck down with a piece of hard plastic. The Positioner is low-cost and durable.