Learn More
Electron Microscopy Sciences EMS Positioning Putty 113 GR
SDP
Catalog No. 501929773
Encompass_Preferred
Blue plastic, non-hardening putty easily positions difficult materials at the angle you want to examine them or holds items in place during specimen preparation. It’s like having an extra hand in the lab! Easily moldable and reusable, the putty will not permanently stick to solid samples or mounts. The blue color offers excellent contrast to most materials under the microscope. Contains odorless acrylic.
Catalog No. 50-192-9773 Supplier Electron Microscopy Sciences Supplier No. 50466
Blue plastic, non-hardening putty easily positions difficult materials at the angle you want to examine them or holds items in place during specimen preparation. It’s like having an extra hand in the lab! Easily moldable and reusable, the putty will not permanently stick to solid samples or mounts. The blue color offers excellent contrast to most materials under the microscope. Contains odorless acrylic.