Learn More
Cayman Chemical PosItIve CNTRL nutralIzng 50uL
SDP
Catalog No. 502894341
Encompass_Preferred
An internal standard for the quantification of heroin by GC- or LC-MS provided as an exempt preparation intended for research and forensic applications
Catalog No. 50-289-4341 Supplier Cayman Chemical Supplier No. 70191650 UL
An internal standard for the quantification of heroin by GC- or LC-MS provided as an exempt preparation intended for research and forensic applications