Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ViewRNA™ Tissue Positive Control Kit (1-plex) (4 rat kidney slides + probe)

Catalog No. QVT0519
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QVT0519 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QVT0519 Supplier Invitrogen™ Supplier No. QVT0519
Add to Cart
Add to Cart

The ViewRNA Tissue Positive Control Kit (1-Plex) is designed for use with the ViewRNA Tissue Assay Core Kit, 1-plex.

The ViewRNA Tissue Positive Control Kit (1-Plex) is designed for use with the ViewRNA Tissue Assay Core Kit, 1-plex (Cat. No. 19931). This positive control kit includes four each of 5-μm thick rat kidney FFPE sections attached to positively charged slides as well as TYPE 1 Rat Ubc Probe Set.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 5 μm thick rat kidney FFPE sections attached to positively charged slides, 4 slides
• TYPE 1 Rat Ubc Probe Set
Format Kit
For Use With (Application) Tissue Analysis
For Use With (Equipment) ViewRNA Tissue Assay Core Kit, 1-plex (Cat. No. 19931)
Product Type RNA In Situ Hybridization Tissue Positive Control Kit
Quantity 1 Kit
Do you offer any positive controls for the ViewRNA tissue assay kits?

Yes, you can use the ViewRNA Tissue Positive Control Kit (1-plex) (Cat. No. QVT0519) or the ViewRNA Tissue Positive Control Kit (2-plex) (4 rat kidney slides + probes) (Cat. No. QVT0516). The positive control includes four each of 5 µm thick rat kidney FFPE sections attached to positively charged slides as well as one (1-plex) or two (2-plex) probe sets for housekeeping genes.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.