Learn More
MEDTOX™ Positive Control for Profile-V MEDTOXScan™ test systems
Catalog No. 22000404
Change view
Click to view available options
Type:
OPI100
OPI2000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-000-404 OPI2000
22-000-403 OPI100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-404 Supplier MEDTOX™ Supplier No. 102365
Add to Cart
Add to Cart
Monitors the performance of Profile-V MEDTOXScan™ drugs of abuse test devices. MEDTOX™ Positive Controls for Profile-V MEDTOXScan test systems are compatible with all quantitative and qualitative drug detection procedures that are sufficiently sensitive to detect the control constituents.
  • Positive control urines have been spiked with authentic reference drug standards and/or appropriate metabolites
  • Should be treated as any “unknown” specimen while following the specific protocol of the assay being used
  • Controls prepared from human based urine available as a negative and Cutoff +50% to monitor the performance of MEDTOX drugs of abuse test devices
  • Each bottle contains stabilized human based urine
Specifications
Sample Type Urine
Type OPI2000
Intended Use Positive control used for quality control with MEDTOXScan drugs of abuse tests
Packaging Bottle
Quantity 5 mL