Learn More
AVIDITY LLC POSITIVE CONTROL PROTEIN SUBST
Catalog No. NC9560198
Encompass
The BIS-300 Kit contains a fully biotinylated MBP-AviTag™ fusion protein standard and an unbiotinylated MBP-AviTag™ fusion protein that may be used as an extent-of-biotinylation comparison for BirA biotin-protein ligase reactions, for SDS-PAGE gel analysis or in Western blot analysis. Kit supplied with the following:  One (1) vial containing 50µL of fully biotinylated MBP-AviTag™ fusion protein at 1mg/mL (50µg).  One (1) vial containing 100µL of unbiotinylated MBP-AviTag™ fusion protein at 1mg/mL (100µg).
Catalog No. NC9560198 Supplier AVIDITY LLC Supplier No. BIS300
The BIS-300 Kit contains a fully biotinylated MBP-AviTag™ fusion protein standard and an unbiotinylated MBP-AviTag™ fusion protein that may be used as an extent-of-biotinylation comparison for BirA biotin-protein ligase reactions, for SDS-PAGE gel analysis or in Western blot analysis. Kit supplied with the following:  One (1) vial containing 50µL of fully biotinylated MBP-AviTag™ fusion protein at 1mg/mL (50µg).  One (1) vial containing 100µL of unbiotinylated MBP-AviTag™ fusion protein at 1mg/mL (100µg).