Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED ON-TARGETplus Non-targeting Control Pool (50 nmol)
Catalog No. NC1269625
Encompass
Negative control pool of four siRNAs designed and microarray tested for minimal targeting of human, mouse or rat genes. ON-TARGETplus modifications reduce potential off-targets. Recommended for determination of baseline cellular responses in RNAi experiments
Catalog No. NC1269625 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. D0018101050
Negative control pool of four siRNAs designed and microarray tested for minimal targeting of human, mouse or rat genes. ON-TARGETplus modifications reduce potential off-targets. Recommended for determination of baseline cellular responses in RNAi experiments