Learn More
Waters Corp Waters Positive Pressure Manifold Spacer, GlycoWorks RapiFluor-MS N-Glycan Kit, 1 pk
Catalog No. 502038293 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This specially designed plastic spacer is intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit workflow when utilizing a positive pressure processor. It is required for proper operation of this workflow with a positive pressure manifold, helping to improve the flow of viscous samples through SPE-packed beds and ensuring highly uniform flow.
  • Specially designed plastic spacer
  • Intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit workflow
  • Required for proper operation with a positive pressure manifold
  • Helps improve the flow of viscous samples through SPE-packed beds
  • Ensures highly uniform flow from well-to-well
Catalog No. 50-203-8293 Supplier Waters Corp Supplier No. 186007987
This specially designed plastic spacer is intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit workflow when utilizing a positive pressure processor. It is required for proper operation of this workflow with a positive pressure manifold, helping to improve the flow of viscous samples through SPE-packed beds and ensuring highly uniform flow.
  • Specially designed plastic spacer
  • Intended for use with the GlycoWorks RapiFluor-MS N-Glycan Kit workflow
  • Required for proper operation with a positive pressure manifold
  • Helps improve the flow of viscous samples through SPE-packed beds
  • Ensures highly uniform flow from well-to-well