Learn More
Post Medical Locking-Top Sharps Container Stand
Catalog No. PM400S
Change view
Click to view available options
Closure Type:
Lockable
Catalog No. Closure Type
PM400S Lockable
1 options
Catalog No. PM400S Supplier Post Medical Supplier No. 400S
Add to Cart
Add to Cart
Holds sharps containers upright and ready for use
  • For use with 1.0, 1.5 and 2.5 gal. Locking-Top Sharps containers
  • Ideal for use on countertops
  • Allows for convenient point-of-use disposal
  • Resist stains and spills
  • Generally used where security is not a concern
Specifications
Closure Type Lockable