Learn More
Aviva Systems Biology Corp POSTN Antibody - N-terminal region (ARP41570_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41570_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POSTN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POSTN. Alternate names: MGC119510, MGC119511, OSF-2, PDLPOSTN, PN, RP11-412K4.1, periostin, OSF2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57908