Learn More
Research Products International Corp Potassium Acetate, 10 KG
SDP
Supplier:  Research Products International Corp P4200010000.0
Encompass_Preferred
In research laboratories, potassium acetate is utilized for various purposes. It is often employed in nucleic acid purification procedures, such as RNA extraction, where it helps precipitate nucleic acids from solution.
Potassium acetate is also used in protein purification techniques, such as ammonium sulfate precipitation, to selectively precipitate proteins from solution based on their solubility characteristics. 
Furthermore, potassium acetate is utilized as a buffering agent in biochemical assays and cell culture media. Its buffering capacity helps maintain a stable pH environment, which is critical for many enzymatic reactions and cell culture processes
Catalog No. 50-223-0498