Learn More
American Radiolabeled Chemicals Inc POT BOROHYDRIDE 100LC
SDP
Catalog No. 50901184
Encompass_Preferred
Potassium borohydride-3H 100mCi as a solid, S.A. 70-100 Ci/mmol, M.W. 53.94, Exclusive
Catalog No. 50-901-184 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1750100MC
Potassium borohydride-3H 100mCi as a solid, S.A. 70-100 Ci/mmol, M.W. 53.94, Exclusive