Learn More
Aqua Solutions Cyanide Standard 1,000 ppm 1 ml = 1 mg CN-, NIST Traceable (250ML)
SDP
Catalog No. NC0469176
Encompass
Cyanide Standard 1,000 ppm 1 ml = 1 mg CN-, NIST Traceable (250ML)
Catalog No. NC0469176 Supplier Aqua Solutions Supplier No. INV001
Cyanide Standard 1,000 ppm 1 ml = 1 mg CN-, NIST Traceable (250ML)