Learn More
Aqua Solutions Aqua Solutions
SDP
Catalog No. NC2066878
Encompass_Preferred
Aqua Solutions is a water filtration system that uses a multi-stage filtration process to remove contaminants from tap water. It features a sediment pre-filter to trap particles, a KDF filter that reduces heavy metals and chlorine, a carbon block filter to remove chemicals and improve taste and odor, and a reverse osmosis membrane to remove dissolved solids. The system connects directly to the cold water line under the sink and has a separate faucet at the sink for dispensing the filtered water. It produces up to 50 gallons of purified water per day while removing up to 99% of contaminants. The Aqua Solutions water filtration system delivers cleaner, tastier water right from the tap.
Catalog No. NC2066878 Supplier Aqua Solutions Supplier No. DOW738
Aqua Solutions is a water filtration system that uses a multi-stage filtration process to remove contaminants from tap water. It features a sediment pre-filter to trap particles, a KDF filter that reduces heavy metals and chlorine, a carbon block filter to remove chemicals and improve taste and odor, and a reverse osmosis membrane to remove dissolved solids. The system connects directly to the cold water line under the sink and has a separate faucet at the sink for dispensing the filtered water. It produces up to 50 gallons of purified water per day while removing up to 99% of contaminants. The Aqua Solutions water filtration system delivers cleaner, tastier water right from the tap.