Learn More
Aqua Solutions POT HYDROX .05N IN METH 1LTR
SDP
Catalog No. NC9302881
Encompass_Preferred
Potassium Hydroxide; 0.05N in Methanol; Clear liquid, methanolic; 1L; M.P. -94 deg.C; B.P. 65 deg.C; Specific Gravity: 0.79; Keep away from heat and flame
Catalog No. NC9302881 Supplier Aqua Solutions Supplier No. 72431L
Potassium Hydroxide; 0.05N in Methanol; Clear liquid, methanolic; 1L; M.P. -94 deg.C; B.P. 65 deg.C; Specific Gravity: 0.79; Keep away from heat and flame