Learn More
Teknova Phosphate Buffer with 0.1% Tween 80. 1000mL, Sterile.
SDP
Catalog No. NC2114479
Encompass
0.778% Sodium Carbonate, 2.622% Monopotassium Phosphate,0.1% Tween-80
This item is not returnable. View return policy
Catalog No. NC2114479 Supplier Teknova Supplier No. P0216
0.778% Sodium Carbonate, 2.622% Monopotassium Phosphate,0.1% Tween-80