Learn More
Aviva Systems Biology Corp POT1 Antibody - middle region (ARP58317_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58317_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POT1. It was validated on Western Blot. POT1,protection of telomeres 1 homolog (S. pombe),HPOT1; DKFZp586D211; POT1,protection of telomeres protein 1,protection of telomeres protein 1,OTTHUMP00000211676,POT1 protection of telomeres 1 homolog,POT1-like telomere end-binding proteinThe immunogen is a synthetic peptide directed towards the middle region of human POT1. Alternate names: DKFZP586D211, hPot1, HPOT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-460