Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium hydroxide, 1310-58-3, MFCD00003553, 1 KG
Catalog No. NC0736931
Encompass
Linear Formula: KOH, Molecular Weight: 56.11, reagent grade, 90%, flakes, Synonym: Caustic potash. Potassium hydroxide (KOH) was used in the following processes: Quantification of glutathione (GSH) and glutathione disulfide (GSSG) in biological samples by HPLC. Preparation of buffer for use in the Investigation of seed fatty acid (FA) composition, protein, oil and sugar content.
Catalog No. NC0736931 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4840161KG
Linear Formula: KOH, Molecular Weight: 56.11, reagent grade, 90%, flakes, Synonym: Caustic potash. Potassium hydroxide (KOH) was used in the following processes: Quantification of glutathione (GSH) and glutathione disulfide (GSSG) in biological samples by HPLC. Preparation of buffer for use in the Investigation of seed fatty acid (FA) composition, protein, oil and sugar content.