Learn More
Potash, sulfurated, S 11.8% min as sulfide
Description
Potash is used in black/brown patina (copper, brass, bronze). Its use include decorative color effects on copper, brass, bronze and nickel. Sulfurated potash has been used in the treatment of acne and other skin disorders usually in the form of a lotion with zinc sulfate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| Physical Form | Chunks |
| Quantity | 100 g |
| Assay Percent Range | S 11.8% min as sulfide |
| Solubility Information | Soluble in water |
| Sensitivity | Air sensitive; Hygroscopic |
| Appearance | Green chunks |
| Chemical Name or Material | Potash, Mixture composed of potassium polysulfides and potassium thiosulfate |
Safety and Handling
| Hazard Category | H228-H314-H335 |
| Precautionary Statement | P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 4.1, Packaging Group: II |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.