Learn More
YSI 3P LifeScience Reference electrolyte, 1 bottle with 250 ml KCl solution 3 mol/l
Catalog No. NC9268020
Encompass_Preferred
Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.
Catalog No. NC9268020 Supplier YSI 3P LifeScience Supplier No. 109705
Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.