Learn More
Medchemexpress LLC Potassium 1,2-naphthoquinone-4-sulphonate | 5908-27-0 | 99.8% | 276.31 | C10H5KO5S | 100mg
SDP
Catalog No. 5000269742
Encompass_Preferred
Potassium 1 2-naphthoquinone-4-sulphonate (Compound 16) is a 6-phosphogluconate dehydrogenase (6PGDH) inhibitor with an IC50 of 53 M Potassium 1 2-naphthoquinone-4-sulphonate can be used in the study of human African trypanosomiasis[1]
Catalog No. 50-002-69742 Supplier Medchemexpress LLC Supplier No. HYW726137100MG
Potassium 1 2-naphthoquinone-4-sulphonate (Compound 16) is a 6-phosphogluconate dehydrogenase (6PGDH) inhibitor with an IC50 of 53 M Potassium 1 2-naphthoquinone-4-sulphonate can be used in the study of human African trypanosomiasis[1]