Learn More
Hach Company Potassium 1 Reagent Powder Pillows, pk/25
Supplier:  Hach Company 1432198
Encompass
For potassium determination in water, soil and plant tissues by the tetraphenylborate method. Hach Method 8049 Sample size: 25 mL. Pack of 25 powder pillows.
Catalog No. 50-199-4996