Learn More
TARGETMOL CHEMICALS INC Potassium 1H-indol-3-yl sulfat
SDP
Catalog No. 502878658
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Potassium 1H-indol-3-yl sulfate (Potassium 3-indoxyl sulfate) is an agonist for the human Aryl Hydrocarbon Receptor (AhR), a key regulator of immune-inflammatory conditions. A metabolite of tryptophan derived from dietary protein, it is produced by intestinal bacteria and metabolized in the liver. In chronic kidney disease patients with impaired renal function, it accumulates in serum as a uremic toxin, inducing oxidative stress and accelerating disease progression. At 250 uM, it activates NF-kb, promoting TGF-beta1 and Smad3 expression in rat proximal tubular cells, which is associated with profibrotic activity. Purity 99.87%
Catalog No. 50-287-8658 Supplier TARGETMOL CHEMICALS INC Supplier No. T493850MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Potassium 1H-indol-3-yl sulfate (Potassium 3-indoxyl sulfate) is an agonist for the human Aryl Hydrocarbon Receptor (AhR), a key regulator of immune-inflammatory conditions. A metabolite of tryptophan derived from dietary protein, it is produced by intestinal bacteria and metabolized in the liver. In chronic kidney disease patients with impaired renal function, it accumulates in serum as a uremic toxin, inducing oxidative stress and accelerating disease progression. At 250 uM, it activates NF-kb, promoting TGF-beta1 and Smad3 expression in rat proximal tubular cells, which is associated with profibrotic activity. Purity 99.87%