Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium 2,4,5-Trichlorobenzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. T062825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T062825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T062825G Supplier TCI America Supplier No. T062825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62625-17-6
Molecular Formula C6H2Cl3KO3S
Molecular Weight (g/mol) 299.587
MDL Number MFCD00024894
InChI Key DRPLRPSXUUSFOB-UHFFFAOYSA-M
Synonym potassium 2,4,5-trichlorobenzenesulfonate, benzenesulfonic acid, 2,4,5-trichloro-, potassium salt, 2,4,5-trichlorobenzenesulfonic acid potassium salt, potassium 2,4,5-trichlorobenzenesulphonate, benzenesulfonic acid, 2,4,5-trichloro-, potassium salt 1:1, potassium 2,4,5-trichlorobenzene-1-sulfonate, 2,4,5-trichlorobenzenesulfonic acid, potassium salt, c6h2cl3so3.k, potassium2,4,5-trichlorobenzenesulfonate, potassium ion 2,4,5-trichlorobenzenesulfonate
PubChem CID 23692292
IUPAC Name potassium;2,4,5-trichlorobenzenesulfonate
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)[O-].[K+]

Specifications

Quantity 25 g
Formula Weight 299.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 2,4,5-Trichlorobenzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.