Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Potassium 2-((6-chloro-dodecafluorohexyl)oxy)-1,1,2,2-tetrafluoroethane-1-sulfonate | 73606-19-6 | MFCD11975311 | ≥98.0% | 570.67 g·mol⁻1 | C8ClF16KO4S | 5 MG
SDP

Catalog No. 5000284116
Encompass_Preferred

Potassium 2-((6-chloro-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexyl)oxy)-1,1,2,2-tetrafluoroethane-1-sulfonate is the potassium salt of a chlorinated perfluoroalkyl ether sulfonate used as a biochemical reagent and reference standard in environmental and toxicology research. It is a persistent fluorinated surfactant with reported reproductive toxicity and endocrine-disrupting effects in biological studies.

  • High purity (≥98.0%).
  • Molecular weight 570.67 g·mol⁻1.
  • Chemical formula C8ClF16KO4S.
  • Supplied as 5 mg solid suitable for analytical and toxicology assays.
  • Reported effects include elevated estradiol and vitellogenin in males and developmental impairment in offspring.
  • Includes SDS and COA for handling and quality documentation.

Catalog No. 50-002-84116 Supplier Medchemexpress LLC Supplier No. HYW5837495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium 2-((6-chloro-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexyl)oxy)-1,1,2,2-tetrafluoroethane-1-sulfonate is the potassium salt of a chlorinated perfluoroalkyl ether sulfonate used as a biochemical reagent and reference standard in environmental and toxicology research. It is a persistent fluorinated surfactant with reported reproductive toxicity and endocrine-disrupting effects in biological studies.

  • High purity (≥98.0%).
  • Molecular weight 570.67 g·mol⁻1.
  • Chemical formula C8ClF16KO4S.
  • Supplied as 5 mg solid suitable for analytical and toxicology assays.
  • Reported effects include elevated estradiol and vitellogenin in males and developmental impairment in offspring.
  • Includes SDS and COA for handling and quality documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.