Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Potassium 2-((6-chloro-dodecafluorohexyl)oxy)-1,1,2,2-tetrafluoroethane-1-sulfonate | 73606-19-6 | MFCD11975311 | >98.0% | 570.67 g/mol | C8ClF16KO4S | 10 MG
SDP

Catalog No. 5000284154
Encompass_Preferred

6:2 Cl-PFAES is a perfluoroalkyl ethanesulfonate potassium salt used in biochemical and toxicology research. The substance is supplied as a high-purity solid and is reported to cause reproductive toxicity, including elevated estradiol and vitellogenin levels and impaired embryonic development. It is intended for laboratory research use only.

  • Chemical formula: C8ClF16KO4S.
  • Molecular weight: 570.67 g/mol.
  • Purity: >98.0%.
  • Appearance: Colorless to off-white solid.
  • Solubility: 100 mg/mL in DMSO; ultrasonic agitation recommended.
  • Storage: Store dry at room temperature; in solution store at -80°C for 2 years or -20°C for 1 year.
  • Intended use: For research use only; not for human therapeutic use.
  • Hazard: Reported reproductive toxicity; handle with appropriate precautions.

Catalog No. 50-002-84154 Supplier Medchemexpress LLC Supplier No. HYW58374910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6:2 Cl-PFAES is a perfluoroalkyl ethanesulfonate potassium salt used in biochemical and toxicology research. The substance is supplied as a high-purity solid and is reported to cause reproductive toxicity, including elevated estradiol and vitellogenin levels and impaired embryonic development. It is intended for laboratory research use only.

  • Chemical formula: C8ClF16KO4S.
  • Molecular weight: 570.67 g/mol.
  • Purity: >98.0%.
  • Appearance: Colorless to off-white solid.
  • Solubility: 100 mg/mL in DMSO; ultrasonic agitation recommended.
  • Storage: Store dry at room temperature; in solution store at -80°C for 2 years or -20°C for 1 year.
  • Intended use: For research use only; not for human therapeutic use.
  • Hazard: Reported reproductive toxicity; handle with appropriate precautions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.