Learn More
Medchemexpress LLC Potassium 2-((6-chloro-dodecafluorohexyl)oxy)-tetrafluoroethane-1-sulfonate | 73606-19-6 | MFCD11975311 | 98.0% | 570.67 g/mol | C8ClF16KO4S | 25 MG
SDP
Catalog No. 5000284238
Encompass_Preferred
6:2 Cl-PFAES (potassium salt) is a chlorinated perfluoroalkyl ethane sulfonate supplied for laboratory research. It is used in environmental, toxicological, and analytical studies of fluorinated surfactants and as a reference standard. The compound is a high-purity material provided in small quantities; consult the safety data sheet before handling due to specific hazard information.
Catalog No. 50-002-84238 Supplier Medchemexpress LLC Supplier No. HYW58374925MG
6:2 Cl-PFAES (potassium salt) is a chlorinated perfluoroalkyl ethane sulfonate supplied for laboratory research. It is used in environmental, toxicological, and analytical studies of fluorinated surfactants and as a reference standard. The compound is a high-purity material provided in small quantities; consult the safety data sheet before handling due to specific hazard information.