Learn More
Hach Company Potassium 2 Reagent Solution Pillows, pk/25
Supplier:  Hach Company 1432298
Encompass
For potassium determination in water, soil and plant tissues by the tetraphenylborate method. Hach Method 8049 Sample size: 25 mL. Pack of 25 powder pillows.
Catalog No. 50-199-4997