Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium (3-Methoxyphenyl)trifluoroborate 98.0+%, TCI America™
SDP

Catalog No. P15805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P15805G 5 g
P15801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P15805G Supplier TCI America Supplier No. P15805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 438553-44-7
Molecular Formula C7H7BF3KO
Molecular Weight (g/mol) 214.036
MDL Number MFCD04039954
InChI Key GKDCWEPJOSDGHY-UHFFFAOYSA-N
Synonym potassium 3-methoxyphenyl trifluoroborate, potassium trifluoro 3-methoxyphenyl borate, potassium 3-methoxyphenyltrifluoroborate, potassium 3-methoxypheny trifluoroborate, potassium trifluoro 3-methoxyphenyl boranuide, amtb102, trifluoro 3-methoxyphenyl potassioboron v, potassium trifluoro-3-methoxyphenyl boranuide, potassium trifluoro 3-methoxyphenyl borate 1-, potassium ion trifluoro 3-methoxyphenyl boranuide
PubChem CID 23697282
IUPAC Name potassium;trifluoro-(3-methoxyphenyl)boranuide
SMILES [B-](C1=CC(=CC=C1)OC)(F)(F)F.[K+]

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Formula Weight 214.04
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Potassium (3-Methoxyphenyl)trifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.