Learn More
Hach Company Potassium 3 Reagent Powder Pillows, pk/100
Supplier:  Hach Company 1432399
Encompass
For potassium determination in water, soil and plant tissues by the tetraphenylborate method. Hach Method 8049 Sample size: 25 mL. Pack of 100 powder pillows.
Catalog No. 50-199-4999