Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium (4-Cyanophenyl)trifluoroborate 95.0+%, TCI America™
SDP

Catalog No. P18155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18155G Supplier TCI America Supplier No. P18155G

missing translation for 'validMainframeMsg'

Specifications

CAS 850623-36-8
Color White
Molecular Formula C7H4BF3KN
MDL Number MFCD04115736
Quantity 5 g
Synonym potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanopheny trifluoroborate, potassium 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyltrifluoroborat, trifluoro 4-cyanophenyl potassioboron v, potassium 4-cyanophenyl-trifluoroboranuide, potassium ion 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoro borate 1-
InChI Key JEQAKQWNUUSKHI-UHFFFAOYSA-N
SMILES [K+].F[B-](F)(F)C1=CC=C(C=C1)C#N
IUPAC Name potassium (4-cyanophenyl)trifluoroboranuide
Molecular Weight (g/mol) 209.02
PubChem CID 23716872
Formula Weight 209.02
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Potassium (4-Cyanophenyl)trifluoroborate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.