Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium 4-Nitrophenyl Sulfate 98.0+%, TCI America™
SDP

Catalog No. N0425200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0425200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0425200MG Supplier TCI America Supplier No. N0425200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6217-68-1
Molecular Formula C6H4KNO6S
Molecular Weight (g/mol) 257.26
MDL Number MFCD00007466
InChI Key BITVAZYUWRLLCN-UHFFFAOYSA-M
Synonym potassium 4-nitrophenyl sulfate, potassium 4-nitrophenyl sulphate, 4-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulfate, sulfuric acid, mono 4-nitrophenyl ester, potassium salt, sulfuric acid, mono 4-nitrophenyl ester, potassium salt 1:1, p-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulphate, phenol, p-nitro-, hydrogen sulfate ester , potassium salt
PubChem CID 80348
IUPAC Name potassium 4-nitrophenyl sulfate
SMILES [K+].[O-][N+](=O)C1=CC=C(OS([O-])(=O)=O)C=C1

Specifications

Melting Point 251°C
Color White-Yellow
Quantity 200 mg
Formula Weight 257.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 4-Nitrophenyl Sulfate
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.