Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium (4-Vinylphenyl)Trifluoroborate

Catalog No. 7235415G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-354-15G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-354-15G Supplier Sigma Aldrich Supplier No. 7235415G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium (4-Vinylphenyl)Trifluoroborate

Specifications

CAS 444910-17-2
Melting Point 331°C
Molecular Formula C54H72Cl4N4Pd2
Linear Formula C54H72Cl4N4Pd2
MDL Number MFCD09039100
Quantity 1 g
Synonym CX 41; Dichloro[1,3-bis(diisopropylphenyl)-2-imidazolidinylidene]palladium(II) dimer; [(SIPr)PdCl2]2
Molecular Weight (g/mol) 1131.83 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.