Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium 6-Hydroxy-2-naphthalenesulfonate 98.0+%, TCI America™
SDP

Catalog No. N070325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N070325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N070325G Supplier TCI America Supplier No. N070325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 833-66-9
Molecular Formula C10H7KO4S
Molecular Weight (g/mol) 262.32
MDL Number MFCD06797149
InChI Key PZWWSVPMGHDZNJ-UHFFFAOYSA-M
Synonym 6-Hydroxy-2-naphthalenesulfonic Acid Potassium Salt, 2-Naphthol-6-sulfonic Acid Potassium Salt, Potassium 2-Naphthol-6-sulfonate, Schaeffer′Cs Acid Potassium Salt
PubChem CID 23677963
IUPAC Name potassium;6-hydroxynaphthalene-2-sulfonate
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)[O-])C=C1O.[K+]

Specifications

Color White
Quantity 25 g
Formula Weight 262.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 6-Hydroxy-2-naphthalenesulfonate
EINECSNumber (4)-1473
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.