Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium AA Standard, 1000 ppm K in 3% HCl, Ricca Chemical
SDP

Catalog No. AK1KH500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AK1KH-500 500 mL Natural Poly Bottle
AK1KH-100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AK1KH-500 Supplier Ricca Chemical Company Supplier No. AK1KH500
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7447-40-7 , 7647-01-0 , 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 95.0, 3.05, 0.16
CAS Max % 99.0, 3.18, 0.17
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Type Standard
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg K (1, 000ppm K)KCl in 3% HCl
Chemical Name or Material Potassium AA Standard
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.