Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acetate, 1 M Solution, pH 7.5, Spectrum™ Chemical
SDP

Catalog No. 18612440 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-440 4 L Amber Glass Bottle
18-612-439 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-612-440 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P17274LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 127-08-2 , 7732-18-5
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.14
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
IUPAC Name potassium acetate
SMILES [K+].CC([O-])=O

Specifications

pH 7.5 ± 0.1
Physical Form Solution
Quantity 4 L
Percent Purity 1.0M ± 0.2M
Grade Ungraded
Concentration or Composition (by Analyte or Components) 1.0 M ± 0.2 M
Packaging Amber Glass Bottle
Chemical Name or Material Potassium Acetate, 1 M Solution, pH 7.5

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.