Learn More
Alkali Scientific Potassium Acetate, 10 Kg
SDP
Catalog No. 5000660409
Encompass_Preferred
Potassium Acetate, Laboratory Grade, is available in a 10 kg package for use in a wide range of laboratory and industrial applications. This salt is commonly used as a buffering agent, especially in biochemical and chemical research. It is effective in maintaining the desired pH in a variety of reactions, making it an essential component in analytical and laboratory procedures. Potassium acetate also finds use in applications involving the preparation of buffer solutions, and it is used in the synthesis of various chemicals. Its high purity ensures that it provides accurate results, making it a preferred choice for precise laboratory experiments and large-scale industrial applications.
Catalog No. 50-006-60409 Supplier Alkali Scientific Supplier No. UMA001CP
Potassium Acetate, Laboratory Grade, is available in a 10 kg package for use in a wide range of laboratory and industrial applications. This salt is commonly used as a buffering agent, especially in biochemical and chemical research. It is effective in maintaining the desired pH in a variety of reactions, making it an essential component in analytical and laboratory procedures. Potassium acetate also finds use in applications involving the preparation of buffer solutions, and it is used in the synthesis of various chemicals. Its high purity ensures that it provides accurate results, making it a preferred choice for precise laboratory experiments and large-scale industrial applications.