Learn More
Biotang Inc POTASSIUM ACETATE 3M 50 ML, MT-243, 3 M Potassium Acetate solution (CH3CO2K). Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50152272 Shop All BIOTANG Inc Products
Encompass_Preferred
POTASSIUM ACETATE 3M 50 ML, MT-243, 3 M Potassium Acetate solution (CH3CO2K). Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-272 Supplier Biotang Inc Supplier No. BTBB819
POTASSIUM ACETATE 3M 50 ML, MT-243, 3 M Potassium Acetate solution (CH3CO2K). Prepared in 18.2 megohms water and filtered through 0.22-micron filter. Shipped at room temperature. Store at room temperature.