Learn More
Alkali Scientific Potassium Acetate, 5 Kilograms
SDP
Catalog No. 5000661024
Encompass_Preferred
Potassium Acetate is a chemical reagent widely used in laboratories for various applications, including biochemical and laboratory research. This 5kg quantity is ideal for high-throughput labs requiring large volumes of a buffer or reagent. Potassium acetate is commonly used in preparing buffer solutions, in the preparation of certain DNA and RNA extraction protocols, and in the production of other chemical reagents. It provides a stable pH environment essential for enzymatic reactions, protein studies, and microbiological analysis. Potassium acetate is frequently used in research involving cell biology, chemical reactions, and other laboratory processes that require precise ionic strength control.
Catalog No. 50-006-61024 Supplier Alkali Scientific Supplier No. UMA005CP
Potassium Acetate is a chemical reagent widely used in laboratories for various applications, including biochemical and laboratory research. This 5kg quantity is ideal for high-throughput labs requiring large volumes of a buffer or reagent. Potassium acetate is commonly used in preparing buffer solutions, in the preparation of certain DNA and RNA extraction protocols, and in the production of other chemical reagents. It provides a stable pH environment essential for enzymatic reactions, protein studies, and microbiological analysis. Potassium acetate is frequently used in research involving cell biology, chemical reactions, and other laboratory processes that require precise ionic strength control.