Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Acetate, 5 M, Ricca Chemical
SDP

Catalog No. R58665001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R58665001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R58665001A Supplier Ricca Chemical Company Supplier No. R58665001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use aqueous solution for laboratory and research applications.

Chemical Identifiers

CAS 7732-18-5 , 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Color Colorless
CAS Min % 54.0, 46.0
CAS Max % 46.0, 54.0
Physical Form Liquid
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 5 Molar
Packaging Natural Poly Bottle
Chemical Name or Material Potassium Acetate, Aqueous
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.